Order Entry
Canada
ContactUsLinkComponent
Cyanide, Complex Certified Reference Standard, SPEX CertiPrep
Cyanide, Complex Certified Reference Standard, SPEX CertiPrep
Catalog # SXRSCN9C-2X
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Cyanide, Complex Certified Reference Standard, SPEX CertiPrep
Catalog # SXRSCN9C-2X
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  RSCN9C-2X
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    2% KOH
  • Pack type:
    LDPE bottle
  • Description:
    Cyanide, Complex Certified Reference Standard
  • Size:
    500 ml
  • Cat. no.:
    SXRSCN9C-2X

Specifications

About this item

Ion Chromarography & Ion Selective Electrode Standards

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers the highest quality IC standards available for the analytical laboratory.