Order Entry
Canada
ContactUsLinkComponent
Ammonium (NH4+) Single-Element Ion Cation Standard, SPEX CertiPrep
Ammonium (NH4+) Single-Element Ion Cation Standard, SPEX CertiPrep
Catalog # SXCS-NH49-2Y
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Ammonium (NH4+) Single-Element Ion Cation Standard, SPEX CertiPrep
Catalog # SXCS-NH49-2Y
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CS-NH49-2Y
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O
  • Pack type:
    LDPE bottle
  • Size:
    125 ml
  • Cat. no.:
    SXCS-NH49-2Y
  • Element:
    Ammonium (NH4+)

Specifications

About this item

Single-element standards for ion chromatography and ion selective electodes.

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary.