Order Entry
Canada
ContactUsLinkComponent
Set of 5 Solutions for Fluoride in H2O, SPEX CertiPrep
Set of 5 Solutions for Fluoride in H2O, SPEX CertiPrep
Catalog # SXAS-F9-SET
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Set of 5 Solutions for Fluoride in H2O, SPEX CertiPrep
Catalog # SXAS-F9-SET
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  AS-F9-SET
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Matrix:
    H₂O
  • Pack type:
    LDPE bottle
  • Description:
    Fluoride solutions, set of 5
  • Size:
    Set of 4x 125 ml standards + 1x 500 ml buffer
  • Cat. no.:
    SXAS-F9-SET

Specifications

About this item

Ion chromatography and ion selective electrode standards.

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers the highest quality IC standards available for the analytical laboratory.