Avantor® Hichrom HI-BIODIESEL 105, GC Columns
Supplier: VWR International
Certificates
About this item
Application specific column for free and total glycerine (phase stable up to 380 °C) and for FAMEs in biodiesel analysis.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76426-554
- 76426-552
- 76426-556
- Phase name:
- HI-BIODIESEL 105
- HI-BIODIESEL 105
- HI-BIODIESEL 105
- Phase composition:
- Proprietary Phase developed for Biodiesel EN14105, D6584 analysis
- Proprietary Phase developed for Biodiesel EN14105 analysis
- Proprietary Phase developed for Biodiesel EN14105 analysis
- Crossbond:
- Yes
- Yes
- Yes
- Temperature limit:
- 380 °C
- 380 °C
- 380 °C