Avantor® Hichrom HI-DEX DAC Beta, GC Columns
Supplier: VWR International
Certificates
About this item
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76429-898
- 76429-896
- 76429-912
- 76429-908
- 76430-062
- 76429-910
- 76430-064
- 76429-904
- 76429-906
- 76429-900
- 76429-902
- Phase name:
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- Phase composition:
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Polarity:
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Crossbond:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Temperature limit:
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C