Avantor® Hichrom HI-PAH, GC Columns
Supplier: VWR International
Certificates
About this item
Application specifc column for polycyclic aromatics hydrocarbons.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76427-132
- 76427-128
- 76427-130
- Phase name:
- HI-PAH
- HI-PAH
- HI-PAH
- Phase composition:
- Proprietary Phase developed for PAHs analysis
- Proprietary Phase developed for PAHs analysis
- Proprietary Phase developed for PAHs analysis
- Crossbond:
- Yes
- Yes
- Yes
- Temperature limit:
- 350 °C
- 350 °C
- 350 °C