ContactUsLinkComponent
Avantor® Hichrom HI-65 HT, GC Columns
Avantor® Hichrom HI-65 HT, GC Columns
Supplier:  VWR International
Certificates
undefined
Avantor® Hichrom HI-65 HT, GC Columns
Supplier:  VWR International
Specifications
  • Catalog No:
  • 76431-674
  • 76431-672
  • 76431-670
  • 76431-668
  • 76431-666
  • Phase name:
  • HI-65 HT
  • HI-65 HT
  • HI-65 HT
  • HI-65 HT
  • HI-65 HT
  • Phase composition:
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • Polarity:
  • Polar stationary phase
  • Polar stationary phase
  • Polar stationary phase
  • Polar stationary phase
  • Polar stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C