Avantor® Hichrom HI-LAP, GC Columns
Supplier: VWR International
Certificates
About this item
Application specific column for lipids, sterols and triglycerides analysis.
Maximum temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76427-122
- Phase name:
- HI-LAP
- Phase composition:
- Proprietary Phase developed for Lipid Analysis Phase, Saturated and Unsaturated Triglycerides, Sterols and Lipid analysis
- Crossbond:
- Yes
- Temperature limit:
- 370 °C