ContactUsLinkComponent
Avantor® Hichrom HI-ALC 1, GC Columns
Avantor® Hichrom HI-ALC 1, GC Columns
Supplier:  VWR International
Certificates
undefined
Avantor® Hichrom HI-ALC 1, GC Columns
Supplier:  VWR International
Specifications
  • Catalog No:
  • 76426-536
  • 76426-534
  • Phase name:
  • HI-ALC 1
  • HI-ALC 1
  • Phase composition:
  • Proprietary Phase developed for blood alcohol analysis
  • Proprietary Phase developed for blood alcohol analysis
  • Crossbond:
  • Yes
  • Yes
  • Temperature limit:
  • 280 °C
  • 280 °C