Order Entry
Canada
ContactUsLinkComponent
Claritas PPT® Grade ICP-MS Spike Sample Standard 1 (Water), SPEX CertiPrep
Claritas PPT® Grade ICP-MS Spike Sample Standard 1 (Water), SPEX CertiPrep
Catalog # SXCL-SPIKE-1
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Claritas PPT® Grade ICP-MS Spike Sample Standard 1 (Water), SPEX CertiPrep
Catalog # SXCL-SPIKE-1
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CL-SPIKE-1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Matrix:
    5% HNO₃/tr. Tartaric acid/tr. HF
  • Pack type:
    LDPE bottle
  • Description:
    ICP-MS Spike Sample Standard 1 (Water)
  • Size:
    125 ml
  • Environmentally Preferable:
  • Cat. no.:
    SXCL-SPIKE-1

Specifications

About this item

as PPT® multi-element spike standards for ICP-MS

SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.

Our Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.

Spike Sample Analysis: Designed for addition to a matrix blank prior to digestion for both water and soil. An aliquot of the respective Spike Standard should be added to produce the proper concentration levels in the digestate.

CLP-M/6020/SW-846 Standards for ICP-MS. The spike standards, CL-SPIKE-1 and CL-SPIKE-2, provide all of the analytes required for ICP-MS. Add 1 mL of CL-SPIKE-1 to aqueous samples and 2 mL of CL-SPIKE-2 to solid samples prior to digestion.

Certifications: SPEX CertiPrep Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.