Order Entry
Canada
ContactUsLinkComponent
Claritas PPT® Grade Zirconium Standard for ICP-MS, SPEX CertiPrep
Claritas PPT® Grade Zirconium Standard for ICP-MS, SPEX CertiPrep
Catalog # SXCLZR2-1BY
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Claritas PPT® Grade Zirconium Standard for ICP-MS, SPEX CertiPrep
Catalog # SXCLZR2-1BY
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CLZR2-1BY
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    LDPE bottle
  • Size:
    125 ml
  • Cat. no.:
    SXCLZR2-1BY
  • Element:
    Zirconium

Specifications

About this item

Single element standard for ICP-MS.

  • Made with an acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)

Claritas PPT® Grade CRMs are designed for ICP and ICP-MS analysis. They are available in single and multi-element solutions. The standards are set at 1 µg/ml, 100 µg/ml, or 1,000 µg/ml and packaged in 30 ml and 125 ml bottles to minimize contamination. They are made using ultra high purity acids, the highest grade starting materials and high purity water in order to minimize contaminants.

Certifications: Certified to DQS to ISO 9001:2015
Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016