Order Entry
Canada
ContactUsLinkComponent
Claritas PPT® Grade ICP-MS Instrument Calibration Standard 2A, SPEX CertiPrep
Claritas PPT® Grade ICP-MS Instrument Calibration Standard 2A, SPEX CertiPrep
Catalog # SXCL-CAL-2A
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Claritas PPT® Grade ICP-MS Instrument Calibration Standard 2A, SPEX CertiPrep
Catalog # SXCL-CAL-2A
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CL-CAL-2A
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃/tr. Tartaric acid
  • Pack type:
    LDPE bottle
  • Description:
    ICP-MS Instrument Calibration Standard 2A
  • Size:
    125 ml
  • Cat. no.:
    SXCL-CAL-2A

Specifications

About this item

Claritas PPT® Multi-Element Standards for ICP-MS. Tuning solutions are for ICP-MS instrumentation tuning and mass calibration prior to analysis.

SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.

The Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.

Instrument Calibration: Dilute to the concentration appropriate for the instrument with equal parts of Claritas PPT® Nitric Acid Blank and Water Blank. For preparation every two weeks, or as needed.

Certifications: SPEX CertiPrep Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.