AvantorĀ® Hichrom HI-FFAP EXT, GC Columns
Supplier: VWR International
Certificates
About this item
Very popular nitroterephthalic-acid-modified polyethylene glycol (PEG) column of high polarity for the analysis of free fatty acids, phenols, alcohols, glycols, with extended temperature range.
- Phase composition: Extended temperature crossbonded nitroterephthalic-acid-modified polyethylene glycol (PEG)
- Long lifetime
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76426-606
- 76426-604
- 76426-840
- 76426-598
- 76426-596
- 76426-838
- 76426-602
- 76426-844
- 76426-600
- 76426-842
- 76426-612
- 76431-306
- 76431-308
- 76426-610
- 76431-302
- 76426-616
- 76431-304
- 76426-614
- 76426-608
- 76426-826
- 76426-824
- 76426-818
- 76426-816
- 76426-822
- 76426-820
- 76426-832
- 76426-590
- 76426-588
- 76426-830
- 76426-594
- 76426-836
- 76426-592
- 76426-834
- 76426-828
- 76426-642
- 76426-640
- 76426-646
- 76426-644
- 76430-558
- 76426-656
- 76426-654
- 76430-554
- 76426-660
- 76430-556
- 76426-658
- 76430-550
- 76426-648
- 76430-552
- 76430-546
- 76426-652
- 76426-650
- 76430-548
- 76430-542
- 76430-544
- 76430-540
- 76426-626
- 76426-620
- 76426-618
- 76426-624
- 76426-622
- 76426-634
- 76426-632
- 76426-638
- 76426-636
- 76426-630
- 76426-628
- 76426-766
- 76426-760
- 76426-758
- 76426-764
- 76426-762
- 76426-686
- 76426-752
- 76426-750
- 76426-684
- 76426-756
- 76426-754
- 76426-700
- 76426-698
- 76426-770
- 76426-704
- 76426-768
- 76426-702
- 76426-692
- 76426-690
- 76426-696
- 76426-694
- 76426-688
- 76426-746
- 76426-744
- 76426-738
- 76426-736
- 76426-742
- 76426-740
- 76426-730
- 76426-664
- 76426-662
- 76426-728
- 76426-734
- 76426-732
- 76426-666
- 76426-678
- 76426-676
- 76426-748
- 76426-682
- 76426-680
- 76430-572
- 76426-670
- 76426-668
- 76426-674
- 76430-568
- 76430-570
- 76426-672
- 76430-564
- 76430-566
- 76430-560
- 76430-562
- 76426-804
- 76426-802
- 76426-806
- 76426-796
- 76426-794
- 76426-800
- 76426-798
- 76426-810
- 76426-808
- 76426-814
- 76426-812
- 76426-782
- 76426-780
- 76426-786
- 76426-784
- 76426-774
- 76426-706
- 76426-772
- 76426-778
- 76426-776
- 76426-788
- 76426-722
- 76426-720
- 76426-792
- 76426-726
- 76426-790
- 76426-724
- 76426-714
- 76426-712
- 76426-718
- 76426-716
- 76426-710
- 76426-708
- Phase name:
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- HI-FFAP EXT
- Phase composition:
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Acid modified Polyethylene Glycol (PEG)
- Polarity:
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- 260 Ā°C
- USP code:
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39
- G14, G15, G16, G25, G35, G39