Order Entry
Canada
ContactUsLinkComponent
Anti-TLR4 Rabbit Polyclonal Antibody
Anti-TLR4 Rabbit Polyclonal Antibody
Certificates
undefined
Anti-TLR4 Rabbit Polyclonal Antibody
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 76309-358
  • 76309-356
  • Antigen symbol:
  • TLR4
  • TLR4
  • Antigen name:
  • TLR4
  • TLR4
  • Antigen synonyms:
  • Rabbit Anti-TLR4 Antibody, ARMD10 antibody, CD284 antibody, Toll (Drosophila) homolog antibody, HToll, Homolog of Drosophila Toll, TLR-4 Antibody, TOLL antibody, Toll Endotoxin Hyporesponsiveness antibody, Toll like receptor 4 antibody, Toll like receptor 4 precursor antibody
  • Rabbit Anti-TLR4 Antibody, ARMD10 antibody, CD284 antibody, Toll (Drosophila) homolog antibody, HToll, Homolog of Drosophila Toll, TLR-4 Antibody, TOLL antibody, Toll Endotoxin Hyporesponsiveness antibody, Toll like receptor 4 antibody, Toll like receptor 4 precursor antibody
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O00206
  • O00206
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Form:
  • Liquid (sterile filtered)
  • Liquid (sterile filtered)
  • Molecular weight:
  • 96
  • 96
  • Purification:
  • Affinity Purified
  • Affinity Purified
  • Storage buffer:
  • 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • Shipping temperature:
  • Dry Ice
  • Dry Ice

Specifications

Related Information

Frequently Bought Together