About this item
PINK1 (PTEN induced putative kinase 1) belongs to the protein kinase superfamily and Ser/Thr protein kinase family. PINK1 protects against mitochondrial dysfunction during cellular stress by phosphorylating mitochondrial proteins. It is involved in the clearance of damaged mitochondria via selective autophagy (mitophagy) by mediating activation and translocation of PARK2. Targets PARK2 to dysfunctional depolarized mitochondria through the phosphorylation of MFN2. Activates PARK2 in 2 steps: (1) by mediating phosphorylation at Ser-65 of PARK2 and (2) mediating phosphorylation of ubiquitin, converting PARK2 to its fully-active form. PINK1 interacts with PARK2 and FBXO7 and forms a complexes with PARK2 and PARK7. PINK1 is associated with Parkinson disease 6 (PARK6). Anti-PINK1 Antibody is useful for researcher interested in neurological research, neurodegenerative disorders, Pink/Parkin Mediated Mitophagy, and Parkinsons Disease Pathway.
Type: Primary
Antigen: PINK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76310-210
- 76310-444
- Antigen symbol:
- PINK1
- PINK1
- Antigen name:
- PINK1
- PINK1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXM7
- Q9BXM7