About this item
p38, also called SAPK2A, belongs to the MAP kinase family. The MAP kinases are critical signaling proteins for many biochemical signals. They participate in transcription regulation, cellular differentiation and proliferation. Environmental signals and cytokines catalyze the kinases into bonding their substrates such as MAX and ATF2, for example, and p53 a tumor suppressing gene. p38 may be associated with diseases such as chlamydia and patellar tendinitis. Anti-p38 antibody is useful for researchers interested in signal pathways, cellular development or cancer research.
Type: Primary
Antigen: MAPK14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76309-336
- 76309-338
- Antigen symbol:
- MAPK14
- MAPK14
- Antigen name:
- P38
- P38
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Goat
- Goat
- Gene ID:
- Q16539
- Q16539