About this item
GRK2 (G Protein-Coupled Receptor Kinase 2) is a member of the G protein-coupled receptor kinase family of proteins. GRK2 Specifically phosphorylates the agonist-occupied form of the beta-adrenergic and as well as a wide range of other substrates including non-GPCR cell surface receptors, and cytoskeletal, mitochondrial, and transcription factor proteins. It is a key regulator of LPAR1 signaling. GRK2 competes with RALA for binding to LPAR1 thus affecting the signaling properties of the receptor. It desensitizes LPAR1 and LPAR2 in a phosphorylation-independent manner. And positively regulates ciliary smoothened (SMO)-dependent Hedgehog (Hh) signaling pathway by facilitating the trafficking of SMO into the cilium and the stimulation of SMO activity. Data from rodent models supports a role for this gene in embryonic development, heart function and metabolism. Elevated expression of this gene has been observed in human patients with heart failure and Alzheimer's disease. Anti-GRK2 Antibody is useful for researchers interested in Neuroscience, Cytokines & Growth Factors, and Stem Cell Research.
Type: Primary
Antigen: GRK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76309-340
- 76309-342
- Antigen symbol:
- GRK2
- GRK2
- Antigen name:
- GRK2
- GRK2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Goat
- Goat
- Gene ID:
- P25098
- P25098