Digital Waving Rotators, Thermo Scientific
Catalog #: 76417-532
Supplier: Thermo Fisher Scientific
Specifications
- Description:Digital waving rotator, US/JP plug
- Max. load:9.8 kg (22 lbs.)
- Control:LED
- Display:LED
- Operating temperature:5...40 °C
- Speed accuracy:±1 rpm ≤30 rpm; ±2 rpm >30 rpm
- Timer:1 min. - 99 hrs. 59 min.
- Weight:15.8 kg (34.8 lbs.)
- Platform material:Stainless steel
- Cat. no.:76417-532
- L×W×H:487×387×354 mm (19.2×15.2×13.9")
- Electrical:120 V, 50/60 Hz, 0.2 A
- Humidity Range:20 - 85% RH
- Speed Range:5 - 120 rpm
- Rotation Angle:1 - 12°
- Platform Dimensions:316×276×19 mm (12.4×10.9×0.7")
Specifications
About this item
Digital Waving Rotator is designed for ELISA, protein synthesis, gel staining and de-staining gels, antibody staining, and other time-sensitive applications.
- Digital display of speed and time
- Platform style: Tray
- Capacity: 6×96-well plates
360° orbital waving rotator for efficient sample mixing with accurate speed control and easy angle adjustment. Optional Non-slip rubber mat tray is designed for carrying plates, dishes, culture bottles and bags.
Certifications: cCSAus, CE, RoHS.
Ordering information: Two year warranty.
Delivery: Includes dimpled rubber mat tray, rubber strip, user manual, EU plug, UK plug, CHN plug, US/Japan plug, replacement rubber tube, screw for rubber strips.
Customers Who Bought This Also Bought