Rhesus macaque Recombinant LAIR-1 (from HEK293)
Catalog # 103739-302
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Rhesus macaque LAIR-1, Mouse IgG2a Fc Tag (Cat. No. LA1-C52A3) with a linear range of 5-78 ng/mL (QC tested).
- Gene ID:ALV66454.1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:LAIR1|CD305
- Protein/peptide name:LAIR-1
- Purity:>95%
- Molecular weight:42.9 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103739-302