Rat Recombinant GITR (from HEK293)
Catalog # 103739-354
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q5M835
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD357|GITR|TNFRSF18|AITR
- Protein/peptide name:GITR
- Purity:>95%
- Molecular weight:12.3 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103739-354