Mouse Recombinant Transferrin (from HEK293)
103737-844
:
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 0.1 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human TfR, His Tag (Cat. No. CD1-H5243) at 2 μg/mL (100 μL/well) can bind Mouse Transferrin, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. TRN-M525b) with a linear range of 0.4-12.5 ng/mL (QC tested).
- Gene ID:Q921I1-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:PRO2086|DKFZp781D0156|PRO1557|Transferrin|TF
- Protein/peptide name:Transferrin
- Purity:>95%
- Molecular weight:101.8 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103737-844