Mouse Recombinant OX40 Ligand (from HEK293)
103738-462
:
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:500 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse OX40 Ligand, His Tag (Cat. No. TN4-M5241) at 0.5 μg/mL (100 μL/well) can bind Mouse OX40, Fc Tag (Cat. No. OX0-M5259) with a linear range of 10-16 ng/mL (QC tested).
- Gene ID:P43488-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TNFSF4|CD134L|TXGP1|OX40L|Glycoprotein Gp34|CD134 ligand|CD252
- Protein/peptide name:OX40 Ligand
- Purity:>90%
- Molecular weight:18.3 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103738-462