Mouse Recombinant CTGF (from HEK293)
Catalog # 103737-812
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in Tris with Potassium glutamate and Arginine, pH7.0. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse CTGF, His Tag (Cat. No. CTF-M52Hc) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CTGF Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (QC tested).; Loaded Monoclonal Anti-Human CTGF Antibody, Human IgG1 on AHC Biosensor, can bind Mouse CTGF, His Tag (Cat. No. CTF-M52Hc) with an affinity constant of 2.46 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P29268-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:IGFBP8|HCS24|IGFBP-8|IBP-8|IGF-binding protein 8|CCN2|NOV2
- Protein/peptide name:CTGF
- Purity:>90%
- Molecular weight:37.3 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Potassium glutamate and Arginine, pH7.0. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103737-812