Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant BCMA (from HEK293)
Mouse Recombinant BCMA (from HEK293)
Catalog # 103739-260
CAS Number:  
Mouse Recombinant BCMA (from HEK293)
Catalog # 103739-260
Supplier Number:  BCAM52H3100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    O88472-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CD269|TNFRSF17|BCM
  • Protein/peptide name:
    BCMA
  • Purity:
    >90%
  • Molecular weight:
    7.3 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT
  • Cat. no.:
    103739-260

Specifications

About this item