Mouse Recombinant B7-H3 (from HEK293)
103738-894
:
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q8VE98
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:B7 homolog 3|B7-H3|CD276
- Protein/peptide name:B7-H3
- Purity:>95%
- Molecular weight:25.4 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103738-894