Human Recombinant PD-L2 (from HEK293)
103739-476
:
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:200 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-PD-L2 MAb (human IgG1) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-L2, Avitag,His Tag (recommended for biopanning) (Cat. No. PD2-H82E8) with a linear range of 0.8-50 ng/mL (QC tested).
- Gene ID:Q9BQ51-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:PDCD1L2|CD273|PD-L2|Butyrophilin B7-DC|PDL2|PDCD1LG2|PDCD1 ligand 2
- Protein/peptide name:PD-L2
- Purity:>95%
- Molecular weight:25.2 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103739-476