Human Recombinant Integrin alpha V beta 5 (from HEK293)
Catalog # 103737-678
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:200 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human Vitronectin at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB5 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT5-H82Wa) with a linear range of 0.01-0.156 μg/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human Fibronectin at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB5 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT5-H82Wa) with a linear range of 0.156-5 μg/mL (Routinely tested).
- Gene ID:P06756-1 (ITGAV) and P18084-1 (ITGB5)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:ITGAV&ITGB5|Integrin alpha V beta 5
- Protein/peptide name:Integrin alpha V beta 5
- Purity:>95%
- Molecular weight:114.7 kDa (ITGAV) and 81.9 kDa (ITGB5)
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103737-678