Human Recombinant Integrin alpha 4 beta 1 (from HEK293)
103737-966
:
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Natalizumab at 1 μg/mL (100 μL/well) can bind Human ITGA4&ITGB1 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT1-H52W1) with a linear range of 1-16 ng/mL (QC tested).
- Gene ID:P13612-1 (ITGA4) and P05556-1 (ITGB1)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Integrin alpha 4 beta 1|ITGA4&ITGB1
- Protein/peptide name:Integrin alpha 4 beta 1
- Purity:>90%
- Molecular weight:111.4 kDa (ITGA4) and 83.7 kDa (ITGB1)
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103737-966