Order Entry
Canada
ContactUsLinkComponent
Human Recombinant IgG1 Fc (from HEK293)
Human Recombinant IgG1 Fc (from HEK293)
Catalog # 103737-768
CAS Number:  
Human Recombinant IgG1 Fc (from HEK293)
Catalog # 103737-768
Supplier Number:  IG1H8213500UG
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    500 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Recombinant Protein G, His Tag (Cat. No. RPG-S3140) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IgG1 Fc, Avitag (Cat. No. IG1-H8213) with a linear range of 0.013-0.512 μg/mL (Routinely tested).; Human FCGRT&B2M Heterodimer Protein, His Tag (SPR and BLI verified) (Cat. No. FCN-H52W7) captured on CM5 Chip via anti-His antibody can bind Biotinylated Human IgG1 Fc, Avitag (Cat. No. IG1-H8213) with an affinity constant of 0.377 μM as determined in SPR assay (Biacore 8K) (Routinely tested).
  • Gene ID:
    P01857-1 
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    IgG1
  • Protein/peptide name:
    IgG1 Fc
  • Purity:
    >95%
  • Molecular weight:
    28.4 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT
  • Cat. no.:
    103737-768

Specifications

About this item