Human Recombinant DNAM-1 (from HEK293)
Catalog # 103738-826
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:200 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Rhesus macaque CD155, His Tag (Cat. No. CD5-R52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human DNAM-1, Fc,Avitag (Cat. No. DN1-H82F9) with a linear range of 10-156 ng/mL (QC tested).; FACS assay shows Biotinylated Human DNAM-1, Fc,Avitag (Cat. No. DN1-H82F9) can bind to 293T cell overexpressing human CD155. The concentration of DNAM-1 is 0.03 μg/mL (Routinely tested).
- Gene ID:Q15762-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD226|PTA1|DNAM1
- Protein/peptide name:DNAM-1
- Purity:>95%
- Molecular weight:54.4 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103738-826