Human Recombinant CD28H (from HEK293)
Catalog # 103738-640
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human B7-H7, His Tag (Cat. No. B77-H52H5) at 5 μg/mL (100 μL/well) can bind Human CD28H, Fc Tag (Cat. No. CDH-H5251) with a linear range of 1-20 ng/mL (QC tested).
- Gene ID:Q96BF3-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TMIGD2|IGPR1
- Protein/peptide name:CD28H
- Purity:>95%
- Molecular weight:40.6 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103738-640