Order Entry
Canada
ContactUsLinkComponent
Human Recombinant CD200 R1 (from HEK293)
Human Recombinant CD200 R1 (from HEK293)
Catalog # 103738-382
CAS Number:  
Human Recombinant CD200 R1 (from HEK293)
Catalog # 103738-382
Supplier Number:  CR2H82F4200UG
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    200 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD200, Fc Tag (Cat. No. OX2-H5251) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD200 R1, Fc,Avitag (Cat. No. CR2-H82F4) with a linear range of 0.5-7.8 ng/mL (QC tested).
  • Gene ID:
    Q8TD46-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    MOX2R|CD200R|OX2R|CRTR2
  • Protein/peptide name:
    CD200 R1
  • Purity:
    >95%
  • Molecular weight:
    55.8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT
  • Cat. no.:
    103738-382

Specifications

About this item