Human Recombinant BAFFR (from HEK293)
Catalog # 103737-694
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 0.01 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 2 μg/mL (100 μL/well) can bind Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (Cat. No. BAR-H5258) with a linear range of 2-63 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (Cat. No. BAR-H5258) at 2 μg/mL (100 μL/well) can bind Biotinylated Human BAFF, His Tag (Cat. No. BAF-H82Q2) with a linear range of 0.2-4 ng/mL (Routinely tested).; Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (Cat. No. BAR-H5258) with an affinity constant of 44.5 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
- Gene ID:Q96RJ3-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CVID4|BAFFR|BAFF-R|BROMIX|CD268|TNFRSF13C|prolixin
- Protein/peptide name:BAFFR
- Purity:>90%
- Molecular weight:34.4 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
- Cat. no.:103737-694