Magid® Clear Anti-Fog PET Full Face Shields with Soft Sponge Padding, Magid Glove
76417-632
- Description:Clear anti-fog PET full face shield with soft sponge padding
- Dimensions:8.6×13"
- Cat. no.:76417-632
Full faceshield features attached soft sponge padding at forehead, elastic band, and removable protective film.
- Clear anti-fog PET full face shield protects the face from liquid splashes
- Attached elastic band removes the need for bulky headgear
- Comfortable soft sponge padding at forehead
- Remove protective film before use
- Caution – strap contains natural rubber latex, which may cause allergic reactions in some individuals
- Not ANSI-rated for impact protection
Designed for use in applications requiring face protection from liquid splashes.
: Appropriate PPE face shield under OSHA 1910.1030 Occupational exposure to bloodborne pathogen and infectious materials.