Order Entry
ContactUsLinkComponent
QuantiChrom™ Biotin Assay Kit, BioAssay Systems
QuantiChrom™ Biotin Assay Kit, BioAssay Systems
Catalog #: 76423-326
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
KIT BIOTIN ASSAY 96/384WELL 100/320TESTS
QuantiChrom™ Biotin Assay Kit, BioAssay Systems
Catalog #: 76423-326
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Biotin assay kit
  • Cat. no.:
    76423-326
  • No. of Tests:
    100 tests

Specifications

About this item

Quantitative determination of biotin in food, cosmetic, and biotinylated proteins.

  • Fast and sensitive: linear detection range - 8 to 200 µm biotin with 30 µl sample (96-well) or 10 to 200 µm biotin with 10 µl sample (384-well)
  • Method: OD 500 nm
  • Samples: food, cosmetics, supplements, and biotinylated proteins or antibodies

BIOTIN , or Vitamin B7, is a water soluble vitamin involved in metabolism, cell growth, and protein synthesis. It is a cofactor to multiple carboxylases necessary for metabolizing fatty acids, glucose, and amino acids. Biotin is found in a wide range of foods and is often taken as a dietary supplement. In the biotechnology industry, biotin is commonly conjugated to proteins in a process called biotinylation. These biotinylated proteins can then be specifically selected using streptavidin and/or avidin's strong affinity for biotin in biochemical assays such as ELISAs.

BioAssay Systems' Biotin Assay Kit is based on avidin's weak affinity for 4'-hydroxyazobenzene-2-carboxylic acid (HABA) and strong affinity for biotin. The avidin-HABA complex reagent has an absorbance at 500nm. When the colored avidin-HABA reagent is introduced to biotin, the biotin binds to avidin, displacing the HABA and causing a decrease in absorbance. The decrease in absorbance at 500nm is directly proportional to the biotin in the sample.

Customers Who Bought This Also Bought