ContactUsLinkComponent
High Temperature Polyester Cleanroom Tape, UltraTape
  89203-840
undefined
High Temperature Polyester Cleanroom Tape, UltraTape
  89203-840
 :  6570CL075P7D
  • Width:
    0.75"
  • Color:
    Clear
  • Length:
    72 yards
  • Material:
    UltraClean
  • Environmentally Preferable:
  • Cat. no.:
    89203-840
  • Supplier No.:
    6570CL075P7D
  • Adhesive material:
    Silicone
Particle-free, residue-free tapes.
  • High temperature clear polyester cleanroom tape
  • Chemical-resistant
  • Autoclavable
Bonds quickly to most surfaces including metal, glass, plastics, and cleanroom paper. Removes cleanly without residue, even after elevated temperature cycles. High temperature masking.