
Hydrochloric acid 0.25 N, Biotech Reagent, J.T.Baker®
Catalog # CAJT0322-09
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
Specifications
- Size:200 L
- Cat. no.:CAJT0322-09
Specifications
Specification Test Results
- Ammonium (NH₄):≤ 3 ppm
- Appearance (Clear, colorless liquid):Passes Test
- Trace Impurities - Heavy Metals:≤ 0.5 ppm
- Normality, Eq/L:0.23 - 0.27
- Sulfate (SO₄):≤ 1 ppm
- Trace Impurities - Iron (Fe):≤ 0.1 ppm
- Endotoxin concentration, EU/mL:≤ 1
Specification Test Results
About this item
:
ADR:8,III
:Management of Change (MOC) category = R
:36.46
UN:1789
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324