Double-Sided Polyester Permanent Adhesion Cleanroom Tape with Liner, UltraTape
Catalog # 89203-824
Supplier: Ultratape Industries
Specifications
- Width:0.75"
- Color:Clear
- Length:36 yards
- Material:UltraClean
- Cat. no.:89203-824
- Supplier No.:1510CL075P3D
- Adhesive material:Acrylic
Specifications
About this item
Particle-free, residue-free tapes.
- Double-sided permanent adhesive tape
Both sides of the polyester carrier have a transparent red Mylar release liner. Ideal for fastening applications, connecting plastic sheeting, or permanent nameplate mounting.