ContactUsLinkComponent
Anti-Static Polyimide Cleanroom Tape, UltraTape
  89203-684
undefined
Anti-Static Polyimide Cleanroom Tape, UltraTape
  89203-684
 :  0157BZ075P3S
  • Width:
    0.75"
  • Color:
    Bronze
  • Length:
    36 yards
  • Material:
    Classic clean
  • Cat. no.:
    89203-684
  • Supplier No.:
    0157BZ075P3S
  • Adhesive material:
    Silicone
Particle-free, residue-free tapes.