Disintegration Tester, Bathless Tablet, sensIR 3200, Distek
Supplier: Avantor
Certificates
About this item
The sensIR 3200 combines two exclusive technologies – bathless heating and Near IR sensing to offer the most innovative disintegration tester. Bathless heating not only reduces warm-up times but eliminates the inconveniences associated with conventional water bath-based instruments.
- Bathless heating
- Auto endpoint detection (optional)
- Run multiple methods simultaneously
- Advanced user interface
- Integrated temperature sensor
- Scalability
- Semi-circle footprint
To match your throughput requirements, the sensIR 3200 is available in two, four or six positions in the smallest foot print of any comparably equipped model.
Ordering information: Does not include installation or qualification services.
Delivery information: Includes baskets, fluted disks, 1 L beakers, and a one-year comprehensive warranty.
Frequently Bought Together