ContactUsLinkComponent
Seals for Captiva ND and ND Lipids Plates, Agilent Technologies
Seals for Captiva ND and ND Lipids Plates, Agilent Technologies
Catalog # CAA8961008
undefined
Seals for Captiva ND and ND Lipids Plates, Agilent Technologies
Catalog # CAA8961008
Supplier Number:  A8961008
Specifications
  • Description:
    DuoSeal 96-well plate seals
  • Cat. no.:
    CAA8961008
Specifications
About this item
Captiva ND enables in-well protein precipitation. Captiva ND Lipids removes biofluid phospholipids.
  • Fully automatable protein precipitation
  • Simple and easy-to-use nondrip protein precipitation plate
  • Resists organic solvent flow until vacuum or positive pressure is applied, allowing for streamlined, fully automatable workflow
  • Dual-depth filter design allows for fast uniform treatment of samples while avoiding membrane plugging or lost samples
  • In-well filter design removes proteins as efficiently as centrifugation while avoiding additional handling steps and accidental pellet transfer
  • Phospholipid removal with Captiva ND Lipids
Captiva ND cartridges and plates are a simple-to-use filtration device designed for high-throughput, automated, in-well protein precipitation. With a unique nondrip (ND) membrane, Captiva ND products enable solvent-first protein precipitation using methanol or acetonitrile. The dual-filter design delivers fast, uniform flow, avoiding filter plugging and ensuring reliable filtrate recovery.;In addition, Captiva ND Lipids cartridges and plates are designed to effectively remove phospholipids from biofluids. The removal of lipid compounds results in enhanced sensitivity, better peak shapes, and more reproducible retention times. Captiva ND and ND Lipids products ensure high sample throughput and help to reduce instrumentation downtime in the laboratory.