Specifications
- Description:Nut and ferrule, stainless steel, ¹/₈", for switching valve
- Cat. no.:AG0100-2637
Specifications
About this item
A range of precision-engineered solvent reservoirs and supplies, designed to provide high levels of sample security and longevity in LC applications.
- Bottles have an ergonomic design, the slim profile and ergonomically shaped gripping zones on both sides of the bottle promote easier and safer use
- A predefined labeling area with self-adhesive labels and colorful silicone bottle tags, mean that you can clearly and unambiguously identify solvent compositions
- An easy to read durable white ceramic print layout clearly indicates the nominal volume of the bottle and allows you to quickly determine the rest contents of the bottle
- Bottles have a GL45 thread and are compatible with stay safe caps
1 and 2 L solvent reservoirs are available in amber and in clear, for situations where sample visibility is important. Bottle head assemblies are available for both tapered and screw-top bottles. The range includes bottle caps with 3-hole inserts, bottle head assemblies with tubing and filters for capillary and nano-systems, and bottle head assemblies with connections for prep systems. Solvent bottles have been redesigned for easier use and safer handling, lowering the risk of accidents in the lab. These redesigned solvent bottles are compatible with your InfinityLab LC series instruments, and can be incorporated seamlessly into your LC workflow.