ContactUsLinkComponent
Spray Chambers and Supplies for PerkinElmer
Spray Chambers and Supplies for PerkinElmer
Catalog # AG8003-0463
New Product
Spray Chambers and Supplies for PerkinElmer
Catalog # AG8003-0463
Supplier Number:  8003-0463
New Product
Specifications
  • Description:
    End cap O-ring, for spray chamber with type A/C/K nebulizers, for Optima 5/7/8×00
  • Environmentally Preferable:
  • Cat. no.:
    AG8003-0463
Specifications
About this item
These spray chambers and supplies are fully compatible with your PerkinElmer ICP-OES.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout. Alternatively, the double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.