OMIX Tips, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Bond Elut OMIX pipette tips provide true high-throughput flexibility for pharmaceutical bioanalysis.
- Fast, uniform flow maximizes productivity and reproducibility
- Small monolithic tip delivers low elution volumes, increasing sensitivity and reducing solvent usage
- Vacuum-free processing improves reproducibility and shortens processing times
Bond Elut OMIX pipette tips reliably purify and enrich femtomole and picomole levels of peptides and proteins prior to MALDI-TOF or LC/MS/MS. This unique monolithic sorbent consistently delivers uniform flow and strong analyte-to-surface interactions. Bond Elut OMIX tips bind more peptides to deliver better sequence coverage.
Frequently Bought Together