Multimode Inlet Supplies for GC, Agilent Technologies
Catalog # CAAGG351060820
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:MultiMode inlet cleaning kit
- Cat. no.:CAAGG351060820
Specifications
About this item
The Multimode Inlet for the Agilent 7890A GC enables improved performance and flexibility in GC and GC/MS laboratories.
- Agilent's premium inlet – two inlets in one for maximum performance and flexibility for the 7890A
Whether you need to increase your system sensitivity with large volume injection capabilities, analyze thermally labile compounds or inject dirty samples, the Multimode Inlet delivers the performance and flexibility you need.