Septa for Headspace Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Frequently Bought Together
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
VWR® Screw-Thread Vials 8-425, Caps and Inserts with Standard Opening
Supplier: VWR International
VWR® Manual Crimping and Decapping Tools
Supplier: VWR International
VWR® TraceClean® Low TOC Vials
Supplier: VWR International
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Supplier: Thermo Scientific
PYREX® Berzelius Beakers, Tall Form, Graduated
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Technologist Choice™ Cotton Tip Applicators, Bio Nuclear Diagnostics
Supplier: Bio Nuclear Diagnostics
Synthetic Chem Elut S Tubes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Standard|Filter Vials
Supplier: THOMSON INSTRUMENT CO
SureSTART™ Glass Snap Top Vials, 2 ml, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
Sandwich Bags, Low Density Flip Top Elkay Plastics
Supplier: Elkay Plastics
Formaldehyde 20% in aqueous solution, EM Grade
Supplier: Electron Microscopy Sciences