Polypropylene Vials, Agilent Technologies
About this item
These vials are appropriate for pH sensitive samples, sodium, and heavy metal analyses.
- Manufactured from virgin PP
- Chemically resistant
Wide opening glass/plastic vials have a glass flanged insert molded to the inside, so sample only comes in contact with high quality glass insert and septum.
Certifications: Meets requirements of 21 CFR 177.1520.
5 Options Available Below
Product Details & Documents
These vials are appropriate for pH sensitive samples, sodium, and heavy metal analyses.
- Manufactured from virgin PP
- Chemically resistant
Wide opening glass/plastic vials have a glass flanged insert molded to the inside, so sample only comes in contact with high quality glass insert and septum.
Certifications: Meets requirements of 21 CFR 177.1520.
Product Family Options
Product Information
- CapacityDiameter ×HMaterialColorCap includedCap SizeAvailabilityPrice
- CA76508-116Crimp/snap top vialsSupplier Number: 5182-0567
- CA5190-3155Crimp/snap top vialsCrimp/snap top vials0.25 ml12×32 mmPPClearNo11 mmDiscontinued-Supplier Number: -
- CA9301-0977Crimp/snap top vials with glass insertCrimp/snap top vials with glass insert0.25 ml12×32 mmPPClearNo11 mmLog in for availabilityPack of 100$333.00Supplier Number: 9301-0977
- CA5188-5390Screw top vials with fixed glass insertScrew top vials with fixed glass insert0.25 ml12×32 mmPPNo12 mmLog in for availabilityPack of 100$344.00Supplier Number: 5188-5390
- CAAG5190-2242Screw top vials, certifiedSupplier Number: 5190-2242
- CAAG5190-2243Screw top vials, certifiedSupplier Number: 5190-2243