Vivacon® 500, Centrifugal Concentrators, Sartorius
Supplier: Sartorius
Certificates
About this item
Vivacon® 500 centrifugal concentrators have been designed for DNA and protein concentration and rebuffering. They feature a horizontal membrane design that is preferred by some users for reasons of consistency with previously used procedures and reproducibility.
- Optimal performance with very dilute samples, e.g. forensic specimens
- Low binding materials; high recoveries even from diluted samples
- Re-spin possibility; complete and highly reproducible sample recovery
- Volume graduation and the molecular weight cut-off printed on individual devices
- For samples up to 500 µl membrane: Regenerated cellulose Hydrosart®
Specifications
- Catalog No:
- CA76408-776
- CA76408-750
- CA76408-772
- CA76408-746
- CA76408-768
- CA76408-744
- CA76408-742
- CA76408-764
- CA76408-784
- CA76408-760
- CA76408-782
- CA76408-780
- Filtration area:
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- 0,32 cm²
- Dead volume:
- 5 µl
- 5 µl
- 5 µl
- 5 µl
- 5 µl
- 5 µl
- 5 µl
- 5 µl
- 5 µl
- 5 µl
- 5 µl
- 5 µl