Order Entry
Canada
ContactUsLinkComponent
PIG® Spill Kit in High-Visibility Storage Chest, NEW PIG
PIG® Spill Kit in High-Visibility Storage Chest, NEW PIG
Catalog # 10820-710
Supplier:  New Pig
undefined
PIG® Spill Kit in High-Visibility Storage Chest, NEW PIG
Catalog # 10820-710
Supplier:  New Pig
Supplier Number:  KIT280-OR

Specifications

  • For Use With:
    Oils, coolants, solvents, water - universal
  • Description:
    PIG® Spill kit in high-visibility storage chest
  • Cat. no.:
    10820-710
  • Absorbency:
    76 gal.

Specifications

About this item

This high capacity, wheeled mobile kit rolls through standard doorways to get you to the spill fast.

  • UV-resistant, lockable polyethylene container keeps absorbents secure and dry indoors or out
  • PIG® Blue socks stop spreading spills, PIG® Mat quickly absorbs, wipers clean surfaces and hands
  • 31"-wide chest fits through standard doors and aisles and has forklift entry for easy moving
  • Highly-visible color helps you locate the kit faster
  • Bold "Spill Kit" lettering for quick identification
  • Mounted on 8" no flat wheels that roll easily over uneven surfaces and brake to keep kit in place

Includes temporary disposal bags, ties and instructions.

Delivery: 10×PIG® Blue absorbent sock with ext. Ø of 3" and 10' length
31×PIG® Blue absorbent sock with ext. Ø of 3" and 48" length
154×PIG® Absorbent mat pad,15 W×20" L
112×PIG® PR40 all-purpose wipers, 15W×20" L
15×Polyethylene disposal bags, 15W×20" L
6×Tamperproof seal label, 2W×6" L
1×Instructions
1×PIG® Storage chest, 31W×48L×41.75" H