About this item
Learn about the effect a catalyst has on a chemical reaction.
- Students will visualize the decomposition of hydrogen peroxide - as the reaction occurs, a large amount of foam is formed that flows up and out of the reaction bottle.
- Aligned to NGSS Practices
- Performance expectations: 2-PS1-4, 2-PS1-3, 5-PS1-1, 5-PS1-4
- Disciplinary core ideas: PS1.A, PS1.B
- Cross cutting concepts include cause and effect, energy and matter, scale proportion and quantity
This activity is a fun, safe way to teach students the effect a catalyst has on a chemical reaction. The decomposition of hydrogen peroxide can be visualized by mixing in a small amount of dish soap and adding yeast as a catalyst. As the reaction occurs, a large amount of foam is formed that flows up and out of the reaction bottle. This kit contains an Instruction Manual and enough materials to perform the demonstration twice.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.